• USA Home
  • I5766 - D-myo-Inositol 4-mono­phos­phate bis(cyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
I5766 Sigma-Aldrich

D-myo-Inositol 4-mono­phos­phate bis(cyclohexylammonium) salt

  • CAS Number 16006-20-5

  • Empirical Formula (Hill Notation) C6H13O9P · 2C6H13N

  • Molecular Weight 458.48

  •  MDL number MFCD00133263

  •  PubChem Substance ID 329815387

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Monosaccharide More...
storage temp.   −20°C
SMILES string   NC1CCCCC1.NC2CCCCC2.O[C@@H]3[C@@H](O)[C@H](O)[C@@H](OP(O)(O)=O)[C@H](O)[C@@H]3O
InChI   1S/2C6H13N.C6H13O9P/c2*7-6-4-2-1-3-5-6;7-1-2(8)4(10)6(5(11)3(1)9)15-16(12,13)14/h2*6H,1-5,7H2;1-11H,(H2,12,13,14)/t;;1-,2-,3-,4+,5-,6-/m..1/s1
InChI key   IMIHZMWNWGIJKQ-PHZSWFKBSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?