• USA Home
  • I6131 - [Ser4, Ile8]-Oxytocin

EMAIL THIS PAGE TO A FRIEND
I6131 Sigma-Aldrich

[Ser4, Ile8]-Oxytocin

≥97% (HPLC)

Synonym: Isotocin, [4-Serine,8-Isoleucine]Oxytocin, [Ser4, Ile8]Oxytocin

  • CAS Number 550-21-0

  • Empirical Formula (Hill Notation) C41H63N11O12S2

  • Molecular Weight 966.14

  •  MDL number MFCD00076734

  •  PubChem Substance ID 24896084

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CCC(C)C(NC(=O)C1CCCN1C(=O)C2CSSCC(N)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(C(C)CC)C(=O)NC(CO)C(=O)NC(CC(N)=O)C(=O)N2)C(=O)NCC(N)=O

Description

Amino Acid Sequence

Cys-Tyr-Ile-Ser-Asn-Cys-Pro-Ile-Gly-NH2 [Disulfide Bridge: 1-6]

Biochem/physiol Actions

Regulates blood pressure.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?