• USA Home
  • I6261 - N4-Isobutyryl-2′-deoxycytidine

EMAIL THIS PAGE TO A FRIEND
I6261 Sigma-Aldrich

N4-Isobutyryl-2′-deoxycytidine

~98%

Purchase

Properties

assay   ~98%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?