EMAIL THIS PAGE TO A FRIEND
I6381 Sigma-Aldrich

β-Ionone

≥95% (GC)

Synonym: 4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

  • CAS Number 14901-07-6

  • Empirical Formula (Hill Notation) C13H20O

  • Molecular Weight 192.30

  •  Beilstein/REAXYS Number 1909544

  •  EC Number 201-224-3

  •  MDL number MFCD00001549

Purchase

Properties

assay   ≥95% (GC)
refractive index   n20/D 1.52 (lit.)
bp   126-128 °C/12 mmHg (lit.)
density   0.945 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   CC(=O)\C=C\C1=C(C)CCCC1(C)C
InChI   1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+
InChI key   PSQYTAPXSHCGMF-BQYQJAHWSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
EN0500000
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

I12603 β-Ionone, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?