• USA Home
  • I7281 - ProteoGel IPG Equilibration Buffer

EMAIL THIS PAGE TO A FRIEND
I7281 Sigma

ProteoGel IPG Equilibration Buffer

Synonym: IPG Equilibration Buffer

Purchase

Description

Application

Following isoelectric focusing, ProteoGel IPG Equilibration Buffer conveniently prepares IPG strips for SDS-PAGE 2D analysis. It adjusts IPG strips to an appropriate pH, maintains protein solubility and further denatures protein samples.

Legal Information

ProteoGel is a trademark of Sigma-Aldrich Co. LLC

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?