• USA Home
  • I7509 - p-Iodo-D-phenylalanine

EMAIL THIS PAGE TO A FRIEND
I7509 Sigma-Aldrich

p-Iodo-D-phenylalanine

Synonym: 2-Amino-3-(4-iodophenyl)-propanoic acid

Purchase

Properties

InChI Key   PZNQZSRPDOEBMS-MRVPVSSYSA-N
mp   251 °C (dec.) (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

294462 p-Iodo-D-phenylalanine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?