• USA Home
  • I7881 - Sodium iminodiacetate dibasic hydrate

EMAIL THIS PAGE TO A FRIEND
I7881 Sigma-Aldrich

Sodium iminodiacetate dibasic hydrate

Synonym: Iminodiacetic acid disodium salt hydrate

  • CAS Number 207398-95-6

  • Linear Formula C4H5NO4Na2 · xH2O

  • Molecular Weight 177.07 (anhydrous basis)

  •  MDL number MFCD00150689

  •  PubChem Substance ID 24896127

Purchase

Properties

SMILES string   O.[Na+].[Na+].[O-]C(=O)CNCC([O-])=O
InChI   1S/C4H7NO4.2Na.H2O/c6-3(7)1-5-2-4(8)9;;;/h5H,1-2H2,(H,6,7)(H,8,9);;;1H2/q;2*+1;/p-2
InChI key   YSBGCHXLMBAKPV-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?