• USA Home
  • I9004 - N-Iodoacetyl-N′-(8-sulfo-1-naphthyl)ethylenediamine

EMAIL THIS PAGE TO A FRIEND
I9004 Sigma-Aldrich

N-Iodoacetyl-N′-(8-sulfo-1-naphthyl)ethylenediamine

crystalline

Synonym: 1,8-I-AEDANS, 8-[2-(2-Iodoacetamido)ethylamino]-1-naphthalenesulfonic acid

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acids, Sulfur Compounds More...
InChI Key   ZDIHEEHLFMMPKM-UHFFFAOYSA-N
form   crystalline
color   yellow
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?