• USA Home
  • K0127 - 15-Ketoprostaglandin F

EMAIL THIS PAGE TO A FRIEND
K0127 Sigma-Aldrich

15-Ketoprostaglandin F

~90%

Synonym: 9,11-Dihydroxy-15-oxo-[5Z,9α,11α,13E]-prosta-5,13-dien-1-oic acid

  • CAS Number 35850-13-6

  • Empirical Formula (Hill Notation) C20H32O5

  • Molecular Weight 352.47

  •  MDL number MFCD00135245

Purchase

Properties

assay   ~90%
shipped in   dry ice
storage temp.   −20°C
SMILES string   CCCCCC(=O)\C=C\[C@H]1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC(O)=O
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?