EMAIL THIS PAGE TO A FRIEND
K1265 Sigma-Aldrich

Ki8751 hydrate

≥98% (HPLC), solid

Synonym: 1-(2,4-Difluoro-phenyl)-3-[4-(6,7-dimethoxy-quinolin-4-yloxy)-2-fluoro-phenyl]-urea hydrate

Purchase

Properties

assay   ≥98% (HPLC)
form   solid
color   white
solubility   DMSO: >20 mg/mL
storage temp.   −20°C

Description

Biochem/physiol Actions

Ki8751 is a potent, selective inhibitor of VEGFR-2 tyrosine kinase (IC50 = 0.9 nM).

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?