EMAIL THIS PAGE TO A FRIEND
K1879 Sigma-Aldrich

Kinetensin

≥97% (HPLC)

  • CAS Number 103131-69-7

  • Linear Formula Ile-Ala-Arg-Arg-His-Pro-Tyr-Phe-Leu

  • Molecular Weight 1172.38

  •  MDL number MFCD00167613

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C

Description

Amino Acid Sequence

Ile-Ala-Arg-Arg-His-Pro-Tyr-Phe-Leu

Biochem/physiol Actions

Neurotensin-related peptide which appears in human plasma treated with pepsin; increases vascular permeability and releases histamine from rat mast cells.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?