• USA Home
  • K1885 - Kinetin hydrochloride

EMAIL THIS PAGE TO A FRIEND
K1885 Sigma-Aldrich

Kinetin hydrochloride

suitable for plant cell culture, ≥96%

Synonym: 6-Furfurylaminopurine, N6-Furfuryladenine

  • CAS Number 177966-68-6

  • Empirical Formula (Hill Notation) C10H9N5O · HCl

  • Molecular Weight 251.67

  •  MDL number MFCD00075757

  •  PubChem Substance ID 24896214

Purchase

Properties

assay   ≥96%
application(s)   cell culture | plant: suitable
solubility   H2O: soluble
storage temp.   −20°C
SMILES string   C(Nc1ncnc2[nH]cnc12)c3ccco3

Description

Application

Kinetin is an adenine-type cytokinin phytohormone that is used in plant culture media such as Murashige & Skoog media in conjunction with auxins such as indole-3-acetic acid (IAA), indole-3-butyric acid (IBA), 1-napthaleneacetic acid (NAA) or others to induce the fomation of callus and to regenerate plant tissues from callus.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Protocols

Growth Regulators - Plant Tissue Culture Protocol

The importance of plant growth regulators in plant tissue culture is well documented. We offer a broad range of plant growth regulators specifically tested for plant cell culture. Each product is ass...
Keywords: Cell culture, Cell division, Culture media, Enzyme activity, Hormones

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?