• USA Home
  • K4378 - Kanamycin sulfate from Streptomyces kanamyceticus

EMAIL THIS PAGE TO A FRIEND
K4378 Sigma-Aldrich

Kanamycin sulfate from Streptomyces kanamyceticus

BioReagent, suitable for plant cell culture

Synonym: Kanamycin A, Kanamycin sulfate salt

Purchase

Properties

product line   BioReagent
potency   ≥750 μg per mg
application(s)   cell culture | plant: suitable
impurities   <5% kanamycin B
Mode of action   protein synthesis | interferes
antibiotic activity spectrum   Gram-negative bacteria
  Gram-positive bacteria
  mycoplasma
SMILES string   OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
InChI   1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1
InChI key   OOYGSFOGFJDDHP-KMCOLRRFSA-N

Description

General description

Chemical structure: aminoglycoside

Packaging

5, 25 g in poly bottle

Application

Used in biotechnology applications to inhibit protein synthesis. Used as a selection agent for cells transformed with kanamycin B (neoR, kanR) resistance gene.

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance:Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Other Notes

This product has been replaced by K1377-SIGMA | Kanamycin sulfate from Streptomyces kanamyceticus powder, BioReagent, suitable for cell culture, suitable for plant cell culture

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
NZ3225030
Protocols & Articles

Protocols

Antibiotics - Plant Tissue Culture Protocol

In general, antibiotics require storage in a refrigerator or freezer. Aminoglycosides (e.g. kanamycin) are hygroscopic and should be stored in a desiccator. Storage of many powdered antibiotics at -2...
Keywords: Adsorption, Antibiotics, Antifungals, Condensations, Molecular biology

Related Content

Antibiotics Selection Guide

The Antibiotic Selection Guide enables researchers to spend more time on their research discoveries and less time seeking established antibiotic information. Link to hundreds of antibiotics for conta...
Keywords: Antibiotics, Cell biology, Cell culture, Genetic, High performance liquid chromatography, Reversed-phase chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?