EMAIL THIS PAGE TO A FRIEND
L0503 Sigma-Aldrich

Leu-Trp

crystalline

  • CAS Number 5156-22-9

  • Empirical Formula (Hill Notation) C17H23N3O3

  • Molecular Weight 317.38

  •  MDL number MFCD00037951

  •  PubChem Substance ID 24896254

Purchase

Properties

form   crystalline
color   light tan
storage temp.   −20°C
SMILES string   CC(C)CC(N)C(=O)NC(Cc1c[nH]c2ccccc12)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?