• USA Home
  • L1017 - trans-6-Leukotriene B4 solution

EMAIL THIS PAGE TO A FRIEND
L1017 Sigma-Aldrich

trans-6-Leukotriene B4 solution

~100 μg/mL in ethanol, ~95%

  • CAS Number 71652-82-9

  • Empirical Formula (Hill Notation) C20H32O4

  • Molecular Weight 336.47

Purchase

Properties

assay   ~95%
concentration   ~100 μg/mL in ethanol
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Packaged under Argon.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?