• USA Home
  • L1625 - DL-Lactic acid hemizinc salt

EMAIL THIS PAGE TO A FRIEND
L1625 Sigma-Aldrich

DL-Lactic acid hemizinc salt

~98%

Purchase

Properties

assay   ~98%
storage temp.   2-8°C

Description

Other Notes

Contains approximately equal amounts of DL isomers.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?