• USA Home
  • L2031 - Leu-Ser-Ser-Phe-Val-Arg-Ile amide acetate salt

EMAIL THIS PAGE TO A FRIEND
L2031 Sigma-Aldrich

Leu-Ser-Ser-Phe-Val-Arg-Ile amide acetate salt

≥97% (HPLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   QTTOOBGNNKXJPC-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?