EMAIL THIS PAGE TO A FRIEND
L2513 Sigma-Aldrich

DL-Lysine

≥98% (TLC)

Synonym: (±)-2,6-Diaminocaproic acid

  • CAS Number 70-54-2

  • Linear Formula NH2(CH2)4CH(NH2)COOH

  • Molecular Weight 146.19

  •  Beilstein/REAXYS Number 1616991

  •  EC Number 200-740-6

  •  MDL number MFCD00064432

  •  PubChem Substance ID 24896317

Purchase

Properties

assay   ≥98% (TLC)
mp   170 °C (dec.) (lit.)
SMILES string   NCCCCC(N)C(O)=O
InChI   1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)
InChI key   KDXKERNSBIXSRK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Amino Acids Reference Charts

Amino acids are the compounds or building blocks that make up peptides and proteins. Each amino acid is structured from an amino group and a carboxyl group bound to a tetrahedral carbon. This carbon ...
Keywords: Building blocks, Gene expression, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?