• USA Home
  • L2636 - [D-pGlu1, D-Phe2, D-Trp3,6]-LH-RH

EMAIL THIS PAGE TO A FRIEND
L2636 Sigma-Aldrich

[D-pGlu1, D-Phe2, D-Trp3,6]-LH-RH

≥97% (HPLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, LH-RH, Peptides and Proteins, Peptides for Cell Biology,
assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@H](CO)NC(=O)[C@@H](Cc4c[nH]c5ccccc45)NC(=O)[C@@H](Cc6ccccc6)NC(=O)[C@H]7CCC(=O)N7)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N8CCC[C@H]8C(=O)NCC(N)=O
InChI   1S/C67H84N16O13/c1-37(2)28-49(59(89)76-48(18-10-26-71-67(69)70)66(96)83-27-11-19-55(83)65(95)74-35-56(68)86)77-62(92)52(31-40-33-72-45-16-8-6-14-43(40)45)80-61(91)51(30-39-20-22-42(85)23-21-39)79-64(94)54(36-84)82-63(93)53(32-41-34-73-46-17-9-7-15-44(41)46)81-60(90)50(29-38-12-4-3-5-13-38)78-58(88)47-24-25-57(87)75-47/h3-9,12-17,20-23,33-34,37,47-55,72-73,84-85H,10-11,18-19,24-32,35-36H2,1-2H3,(H2,68,86)(H,74,95)(H,75,87)(H,76,89)(H,77,92)(H,78,88)(H,79,94)(H,80,91)(H,81,90)(H,82,93)(H4,69,70,71)/t47-,48+,49+,50-,51+,52-,53-,54+,55+/m1/s1
InChI key   NYWIVATZZKIKCF-FKDXDCHSSA-N

Description

Amino Acid Sequence

D-pGlu-D-Phe-D-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-Gly-NH2

Biochem/physiol Actions

Substance P antagonist; inhibits spinal cord vasomotor responses

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?