EMAIL THIS PAGE TO A FRIEND
L2751 Sigma-Aldrich

Linolenyl alcohol

≥99%, liquid

Synonym: 9,12,15-Octadecatrienol

  • CAS Number 506-44-5

  • Empirical Formula (Hill Notation) C18H32O

  • Molecular Weight 264.45

  •  EC Number 208-039-7

Purchase

Properties

assay   ≥99%
form   liquid
storage temp.   −20°C
InChI   1S/C18H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h3-4,6-7,9-10,19H,2,5,8,11-18H2,1H3/b4-3-,7-6-,10-9-
InChI key   IKYKEVDKGZYRMQ-PDBXOOCHSA-N

Description

Packaging

Sealed ampule.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?