EMAIL THIS PAGE TO A FRIEND
L3915 Sigma-Aldrich

Leu-Ala hydrate

vial of 2 mg

Synonym: L-Leucyl-L-alanine hydrate

  • CAS Number 7298-84-2 (anhydrous)

  • Linear Formula (CH3)2CHCH2CH(NH2)CONHCH(CH3)COOH · xH2O

  • Molecular Weight 202.25 (anhydrous basis)

  •  Beilstein/REAXYS Number 1726165

  •  MDL number MFCD00149112

  •  PubChem Substance ID 24896356

Purchase

Properties

packaging   vial of 2 mg
storage temp.   2-8°C
SMILES string   O.CC(C)C[C@H](N)C(=O)N[C@@H](C)C(O)=O
InChI   1S/C9H18N2O3.H2O/c1-5(2)4-7(10)8(12)11-6(3)9(13)14;/h5-7H,4,10H2,1-3H3,(H,11,12)(H,13,14);1H2/t6-,7-;/m0./s1
InChI key   FTDFAGANYJVHDA-LEUCUCNGSA-N

Description

Application

Enhances the fluorescence in phenylalanine-ninhydrin reactions.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?