EMAIL THIS PAGE TO A FRIEND
L4752 Sigma-Aldrich

DL-Leu-DL-Phe

  • CAS Number 56217-82-4

  • Empirical Formula (Hill Notation) C15H22N2O3

  • Molecular Weight 278.35

  •  MDL number MFCD00037258

  •  PubChem Substance ID 24896378

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)CC(N)C(=O)NC(Cc1ccccc1)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?