• USA Home
  • L5629 - 1-Dodecanoyl-sn-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
L5629 Sigma-Aldrich

1-Dodecanoyl-sn-glycero-3-phosphocholine

~99%

Synonym: L-α-Lysophosphatidylcholine, lauroyl, Lysolecithin, lauroyl

  • CAS Number 20559-18-6

  • Empirical Formula (Hill Notation) C20H42NO7P

  • Molecular Weight 439.52

  •  MDL number MFCD00133434

  •  PubChem Substance ID 24896409

Purchase
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?