EMAIL THIS PAGE TO A FRIEND
L6378 Sigma-Aldrich

Ethyl γ-linolenate

≥99%, liquid

Synonym: γ-Linolenic acid ethyl ester, 6,9,12-Octadecatrienoic acid ethyl ester

Purchase

Properties

assay   ≥99%
form   liquid
storage temp.   −20°C
SMILES string   CCCCC\C=C/C\C=C/C\C=C/CCCCC(=O)OCC

Description

Packaging

Sealed ampule.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?