• USA Home
  • L6401 - Lacto-N-fucopentaose II

EMAIL THIS PAGE TO A FRIEND
L6401 Sigma

Lacto-N-fucopentaose II

from human milk, ≥98% (HPAE/PAD)

Synonym: β-D-Gal-(1→3)-(α-L-Fuc-[1→4])-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-D-Glc, LNFP II, Lea-Lactose, Lewis-a pentasaccharide

  • CAS Number 21973-23-9

  • Empirical Formula (Hill Notation) C32H55NO25

  • Molecular Weight 853.77

  •  Beilstein Registry Number 6050703

  •  MDL number MFCD00171398

Purchase

Properties

biological source   human milk
assay   ≥98% (HPAE/PAD)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?