EMAIL THIS PAGE TO A FRIEND
L7154 Sigma-Aldrich

[Hyp9]-LH-RH

≥97% (HPLC)

Synonym: pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Hyp-Gly-NH2

  • CAS Number 67019-13-0

  • Empirical Formula (Hill Notation) C55H75N17O14

  • Molecular Weight 1198.29

  •  MDL number MFCD00214314

  •  PubChem Substance ID 24896454

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, LH-RH, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   IFKVSSKFSAFSGI-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Nts(299757)

Description

Amino Acid Sequence

pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Hyp-Gly-NH2

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?