• USA Home
  • L7279 - [Gly6]-Locustapyrokinin

EMAIL THIS PAGE TO A FRIEND
L7279 Sigma-Aldrich

[Gly6]-Locustapyrokinin

Synonym: pGlu-Asp-Ser-Gly-Asp-Gly-Trp-Pro-Gln-Gln-Pro-Phe-Val-Pro-Arg-Leu-NH2

  • CAS Number 152575-14-9

  • Empirical Formula (Hill Notation) C82H117N23O24

  • Molecular Weight 1808.95

  •  MDL number MFCD00214214

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]1CCCN1C(=O)[C@@H](NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]4CCCN4C(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)CNC(=O)[C@H](CC(O)=O)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H]7CCC(=O)N7)C(C)C)C(N)=O
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?