EMAIL THIS PAGE TO A FRIEND
L7404 Sigma-Aldrich

Histatin 8

≥95% (HPLC)

Synonym: Lys-Phe-His-Glu-Lys-His-His-Ser-His-Arg-Gly-Tyr

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   JQZXHRXVJFXBKA-UHFFFAOYSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Hemagglutination inhibiting peptide originally isolated from human parotid saliva

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?