EMAIL THIS PAGE TO A FRIEND
L7750 Sigma-Aldrich

D-Leucine

≥98% (TLC)

Synonym: (R)-2-Amino-4-methylpentanoic acid

  • CAS Number 328-38-1

  • Linear Formula (CH3)2CHCH2CH(NH2)CO2H

  • Molecular Weight 131.17

  •  Beilstein/REAXYS Number 1721721

  •  EC Number 206-327-7

  •  MDL number MFCD00063088

Purchase

Properties

assay   ≥98% (TLC)
mp   >300 °C (lit.)
SMILES string   CC(C)C[C@@H](N)C(O)=O
InChI   1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m1/s1
InChI key   ROHFNLRQFUQHCH-RXMQYKEDSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

855448 D-Leucine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?