• USA Home
  • L7902 - Lacto-N-fucopentaose V

EMAIL THIS PAGE TO A FRIEND
L7902 Sigma-Aldrich

Lacto-N-fucopentaose V

from human milk, ≥95%

Synonym: β-D-Gal-(1→3)-β-D-GlcNAc-(1→3)-β-D-Gal-(1→4)-[α-L-Fuc-(1→3)]-D-Glc, LNFP V

  • CAS Number 60254-64-0

  • Empirical Formula (Hill Notation) C32H55NO25

  • Molecular Weight 853.77

  •  MDL number MFCD00214317

  •  PubChem Substance ID 24896487

Purchase

Properties

biological source   human milk
assay   ≥95%
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?