• USA Home
  • L8139 - Lys-Ala-7-amido-4-methylcoumarin dihydrochloride

EMAIL THIS PAGE TO A FRIEND
L8139 Sigma-Aldrich

Lys-Ala-7-amido-4-methylcoumarin dihydrochloride

  • CAS Number 103404-62-2

  • Empirical Formula (Hill Notation) C19H26N4O4 · 2HCl

  • Molecular Weight 447.36

  •  MDL number MFCD00058180

  •  PubChem Substance ID 24896494

Purchase

Description

Substrates

Substrate for rat brain dipeptidyl aminopeptidase

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?