• USA Home
  • L8158 - Lithium sulfate monohydrate

EMAIL THIS PAGE TO A FRIEND
L8158 Sigma-Aldrich

Lithium sulfate monohydrate

BioXtra, ≥99.0% (titration)

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Lithium, Molecular Biology, Optimization Reagents,
product line   BioXtra
assay   ≥99.0% (titration)
impurities   <0.0005% Phosphorus (P)
  <0.01% Insoluble matter
solubility   H2O: 1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): <0.002%
cation traces   Al: <0.0005%
  Ca: <0.0005%
  Cu: <0.0005%
  Fe: <0.0005%
  K: <0.05%
  Mg: <0.0005%
  NH4+: <0.05%
  Na: <0.05%
  Pb: <0.001%
  Zn: <0.0005%
SMILES string   [Li+].[Li+].O.[O-]S([O-])(=O)=O
InChI   1S/2Li.H2O4S.H2O/c;;1-5(2,3)4;/h;;(H2,1,2,3,4);1H2/q2*+1;;/p-2
InChI key   RKGLUDFWIKNKMX-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?