EMAIL THIS PAGE TO A FRIEND
L8632 Sigma-Aldrich

DL-Leu-DL-Tyr

  • CAS Number 19659-00-8

  • Empirical Formula (Hill Notation) C15H22N2O4

  • Molecular Weight 294.35

  •  MDL number MFCD00063033

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)CC(N)C(=O)NC(Cc1ccc(O)cc1)C(O)=O

Description

Other Notes

Although our supplier claims this is an equal mixture of the four diastereomers, our assay indicates that the composition varies from lot to lot.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?