EMAIL THIS PAGE TO A FRIEND
L8641 Sigma-Aldrich

Palmityl dodecanoate

~99%

Synonym: Dodecanoic acid hexadecanyl ester, Lauric acid palmityl ester, Palmityl laurate

Purchase

Properties

assay   ~99%
form   solid
functional group   ester
shipped in   ambient
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCC
InChI   1S/C28H56O2/c1-3-5-7-9-11-13-14-15-16-17-19-21-23-25-27-30-28(29)26-24-22-20-18-12-10-8-6-4-2/h3-27H2,1-2H3
InChI key   VWOKINHIVGKNRX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?