EMAIL THIS PAGE TO A FRIEND
L9539 Sigma-Aldrich

L-655,240

≥98% (HPLC), solid

Synonym: 3-[1-(4-Chlorobenzyl)-5-fluoro-3-methyl-indol-2-yl]-2,2-dimethyl propanoic acid

Purchase

Properties

assay   ≥98% (HPLC)
form   solid
color   off-white
solubility   DMSO: 30 mg/mL
  H2O: insoluble
Gene Information   human ... TBXA2R(6915)

Description

Biochem/physiol Actions

Thromboxane A2 receptor antagonist, induces platelet aggregation and contraction of vascular and smooth muscle. Increases PI turnover with elevation of intracellular free Ca2+.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?