• USA Home
  • M0656 - 1-O-Methyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
M0656 Sigma

1-O-Methyl-rac-glycerol

Synonym: (±)-3-Methoxy-1,2-propanediol

  • CAS Number 36887-04-4

  • Empirical Formula (Hill Notation) C4H10O3

  • Molecular Weight 106.12

  •  MDL number MFCD00004719

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

260401 3-Methoxy-1,2-propanediol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?