• USA Home
  • M115 - (±)-Methoxyverapamil hydrochloride

EMAIL THIS PAGE TO A FRIEND
M115 Sigma-Aldrich

(±)-Methoxyverapamil hydrochloride

Synonym: (±)-α-[3-[(2-(3,4-Dimethoxyphenyl)ethyl)methylamino]propyl]-3,4,5-trimethoxy-α-(1-methylethyl)benzeneacetonitrile hydrochloride, D 600, Gallopamil

  • CAS Number 16662-47-8

  • Empirical Formula (Hill Notation) C28H40N2O5 · HCl

  • Molecular Weight 521.09

  •  MDL number MFCD00058000

Purchase

Properties

application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Veterinary
SMILES string   Cl[H].COc1ccc(CCN(C)CCCC(C#N)(C(C)C)c2cc(OC)c(OC)c(OC)c2)cc1OC
InChI   1S/C28H40N2O5.ClH/c1-20(2)28(19-29,22-17-25(33-6)27(35-8)26(18-22)34-7)13-9-14-30(3)15-12-21-10-11-23(31-4)24(16-21)32-5;/h10-11,16-18,20H,9,12-15H2,1-8H3;1H
InChI key   OKCRIUNHEQSXFD-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Blocks L-type Ca2+ channels

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M5644 (±)-Methoxyverapamil hydrochloride, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?