• USA Home
  • M139 - S(+)-3,4-Methylenedioxymethamphetamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
M139 Sigma-Aldrich

S(+)-3,4-Methylenedioxymethamphetamine hydrochloride

solid

Synonym: Ecstasy, S-(+)-MDMA hydrochloride

  • CAS Number 64057-70-1

  • Empirical Formula (Hill Notation) C11H15NO2 · HCl

  • Molecular Weight 229.70

  •  MDL number MFCD00153834

  •  PubChem Substance ID 24896642

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, M,
InChI Key   LUWHVONVCYWRMZ-QRPNPIFTSA-N
form   solid
optical activity   [α]24/D +18.38°, c = 1 in H2O(lit.)
drug control   USDEA Schedule I; Home Office Schedule 1; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)
color   white
solubility   H2O: soluble

Description

Biochem/physiol Actions

Hallucinogen; releases dopamine and serotonin from nerve terminals; selective serotonergic neurotoxin.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
SH5700000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?