EMAIL THIS PAGE TO A FRIEND
M1770 Sigma-Aldrich

Methanol

BioReagent, suitable for protein sequencing

Synonym: Methyl alcohol

Purchase

Properties

Related Categories Apoptosis and Cell Cycle, Cell Biology, Cell Signaling and Neuroscience, Core Bioreagents, General Reagents,
vapor density   1.11 (vs air)
vapor pressure   410 mmHg ( 50 °C)
  97.68 mmHg ( 20 °C)
product line   BioReagent
assay   ≥99.9%
autoignition temp.   725 °F
expl. lim.   36 %
refractive index   n20/D 1.329 (lit.)
bp   64.7 °C (lit.)
mp   −98 °C (lit.)
density   0.791 g/mL at 25 °C (lit.)
suitability   suitable for protein sequencing
SMILES string   CO
InChI   1S/CH4O/c1-2/h2H,1H3
InChI key   OKKJLVBELUTLKV-UHFFFAOYSA-N

Description

Other Notes

Protein sequencing grade

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
RTECS 
PC1400000
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Single- & multi-channel pipettes from BRAND
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?