• USA Home
  • M2251 - 5-Methoxy-3-indoleacetic acid

EMAIL THIS PAGE TO A FRIEND
M2251 Sigma-Aldrich

5-Methoxy-3-indoleacetic acid

crystalline

Synonym: 2-(5-Methoxy-3-indolyl)acetic acid

  • CAS Number 3471-31-6

  • Empirical Formula (Hill Notation) C11H11NO3

  • Molecular Weight 205.21

  •  Beilstein/REAXYS Number 187161

  •  EC Number 222-438-3

  •  MDL number MFCD00005638

Purchase

Properties

InChI Key   COCNDHOPIHDTHK-UHFFFAOYSA-N
form   crystalline
color   light yellow to tan
mp   145-148 °C (dec.) (lit.)
storage temp.   2-8°C

Description

Application

• Reactant for preparation of aryloxybenzothiazoles as inhibitors of NO production
• Reactant for preparation of transcription inhibitors of novel Gli1 as antitumor agents
• Reactant for preparation of indole derivatives as NR2B/NMDA receptor antagonists
• Reactant for preparation of indolyl esters and amides related to indomethacin as selective COX-2 inhibitors
• Reactant for preparation of quinoline salicylic acid series of P-selectin antagonists
• Reactant for preparation of prostaglandin D2 receptor antagonists

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M14935 5-Methoxy-3-indoleacetic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?