• USA Home
  • M2282 - Malformin A from Aspergillus niger

EMAIL THIS PAGE TO A FRIEND
M2282 Sigma-Aldrich

Malformin A from Aspergillus niger

~95% (TLC)

  • CAS Number 3022-92-2

  • Empirical Formula (Hill Notation) C23H39N5O5S2

  • Molecular Weight 529.72

Purchase

Properties

assay   ~95% (TLC)
storage temp.   2-8°C

Description

Biochem/physiol Actions

A cyclic pentapeptide antibiotic which causes malformed roots in cereals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ON7050000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?