• USA Home
  • M2387 - Methyl 3-amino-3-deoxy-β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
M2387 Sigma-Aldrich

Methyl 3-amino-3-deoxy-β-D-glucopyranoside

  • CAS Number 14133-36-9

  • Empirical Formula (Hill Notation) C7H15NO5

  • Molecular Weight 193.20

  •  MDL number MFCD00047530

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?