• USA Home
  • M2897 - N-Methoxysuccinyl-Ala-Ala-Pro-Val

EMAIL THIS PAGE TO A FRIEND
M2897 Sigma-Aldrich

N-Methoxysuccinyl-Ala-Ala-Pro-Val

Purchase

Properties

storage temp.   −20°C
SMILES string   COC(=O)CCC(=O)NC(C)C(=O)NC(C)C(=O)N1CCCC1C(=O)NC(C(C)C)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?