• USA Home
  • M3393 - Methaqualone hydrochloride

EMAIL THIS PAGE TO A FRIEND
M3393 Sigma-Aldrich

Methaqualone hydrochloride

1.0 mg/mL in methanol

Synonym: 2-Methyl-3-(2-methylphenyl)-4(3H)-quinazolinone hydrochloride

Purchase

Properties

drug control   USDEA Schedule I; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
storage temp.   2-8°C
SMILES string   Cl.Cc1ccccc1N2C(C)=Nc3ccccc3C2=O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?