EMAIL THIS PAGE TO A FRIEND
M3443 Sigma-Aldrich

MSX-2

≥98% (HPLC), solid

Synonym: 3-(3-Hydroxypropyl)-8-(m-methoxystyryl)-7-methyl-1-propargylxanthine

  • CAS Number 261717-18-4

  • Empirical Formula (Hill Notation) C21H22N4O4

  • Molecular Weight 394.42

  •  MDL number MFCD11046007

Purchase

Properties

assay   ≥98% (HPLC)
form   solid
color   pale yellow
solubility   DMSO: ~14 mg/mL
  H2O: insoluble
storage temp.   −20°C
SMILES string   COc1cccc(\C=C\c2nc3N(CCCO)C(=O)N(CC#C)C(=O)c3n2C)c1

Description

Biochem/physiol Actions

Selective A2A adenosine receptor antagonist

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?