• USA Home
  • M4018 - DL-α-Neopentylglycine

EMAIL THIS PAGE TO A FRIEND
M4018 Sigma-Aldrich

DL-α-Neopentylglycine

Synonym: β-tert-Butyl-DL-alanine, γ-Methyl-DL-leucine, (±)-2-Amino-4,4-dimethylpentanoic acid

  • CAS Number 106247-35-2

  • Linear Formula (CH3)3CCH2CH(NH2)COOH

  • Molecular Weight 145.20

  •  Beilstein/REAXYS Number 3931664

  •  MDL number MFCD00066078

Purchase

Properties

mp   272-274 °C (dec.)
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?