• USA Home
  • M4068 - Mesoridazine benzenesulfonate

EMAIL THIS PAGE TO A FRIEND
M4068 Sigma-Aldrich

Mesoridazine benzenesulfonate

≥98% (HPLC), powder

Synonym: 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine

Purchase

Properties

Related Categories Antagonists - Dopaminergics, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Biochemicals and Reagents, Cell Biology,
assay   ≥98% (HPLC)
form   powder
color   white
solubility   DMSO: ≥10 mg/mL
storage temp.   2-8°C
SMILES string   OS(=O)(=O)c1ccccc1.CN2CCCCC2CCN3c4ccccc4Sc5ccc(cc35)S(C)=O
InChI   1S/C21H26N2OS2.C6H6O3S/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(26(2)24)15-19(21)23;7-10(8,9)6-4-2-1-3-5-6/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1-5H,(H,7,8,9)
InChI key   CRJHBCPQHRVYBS-UHFFFAOYSA-N
Gene Information   human ... DRD2(1813), HTR2A(3356)

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

Mesoridazine benzenesulfonate is a D2 dopamine receptor antagonist; antipsychotic.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DB7176700

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

UHPLC Analysis of Mesoridazine and Haloperidol on Titan™ C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Mesoridazine and Haloperidol on Titan™ C18
Keywords: Chromatography, Mass spectrometry, Pharmaceutical

UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?