• USA Home
  • M4293 - 7-Methoxycoumarin-4-acetyl-Pro-Leu-Gly

EMAIL THIS PAGE TO A FRIEND
M4293 Sigma-Aldrich

7-Methoxycoumarin-4-acetyl-Pro-Leu-Gly

Synonym: MOCAc-Pro-Leu-Gly

  • CAS Number 140430-56-4

  • Empirical Formula (Hill Notation) C25H31N3O8

  • Molecular Weight 501.53

  •  MDL number MFCD00210686

Purchase

Properties

storage temp.   −20°C

Description

Application

Fluorescent peptide for use as a reference.

Other Notes

The product of the action of matrix metalloproteinases on the internally quenched substrate M6412.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?