• USA Home
  • M4400 - Methyl Red hydrochloride

EMAIL THIS PAGE TO A FRIEND
M4400 Sigma-Aldrich

Methyl Red hydrochloride

ACS reagent

Synonym: 4-Dimethylaminoazobenzene-2′-carboxylic acid hydrochloride, Acid Red 2

  • CAS Number 63451-28-5

  • Linear Formula (CH3)2NC6H4N=NC6H4CO2H·HCl

  • Molecular Weight 305.76

  •  Colour Index Number 13020

  •  EC Number 264-190-9

  •  MDL number MFCD00012614

Purchase

Properties

grade   ACS reagent
visual transition interval   4.2 - 6.2, pink to yellow
mp   175 °C (dec.) (lit.)
SMILES string   Cl.CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O
InChI   1S/C15H15N3O2.ClH/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);1H/b17-16+;
InChI key   ICTWKXAVJAHRMK-CMBBICFISA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?