• USA Home
  • M4418 - 4-Methyl-3-phenylcoumarin

EMAIL THIS PAGE TO A FRIEND
M4418 Sigma-Aldrich

4-Methyl-3-phenylcoumarin

98% (HPLC)

  • CAS Number 23028-23-1

  • Empirical Formula (Hill Notation) C16H12O2

  • Molecular Weight 236.27

  •  MDL number MFCD00192313

Purchase

Properties

assay   98% (HPLC)
mp   156-158 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?